HEPES Buffer, 1M Solution, pH 7.3 (Molecular Biology), Fisher BioReagents™
HEPES Buffer, 1M Solution, pH 7.3 (Molecular Biology), Fisher BioReagents™
HEPES Buffer, 1M Solution, pH 7.3 (Molecular Biology), Fisher BioReagents™
HEPES Buffer, 1M Solution, pH 7.3 (Molecular Biology), Fisher BioReagents™
HEPES Buffer, 1M Solution, pH 7.3 (Molecular Biology), Fisher BioReagents™
Fisher Chemical
HEPES Buffer, 1M Solution, pH 7.3 (Molecular Biology), Fisher BioReagents™
Commonly used buffering agent
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
1 L
Packaging:
View all
Poly Bottle
Catalog number FBRBP299-500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS7365-45-9, 7732-18-5
Specifications
Absorbance≤0.01(0.1M solution) at 260nm, 0.004 max. (0.1M solution) at 280nm
Chemical Name or MaterialHEPES Buffer
Name Note1M Solution, pH 7.3
CAS7365-45-9
ChemAlert Storage SymbolGray
View more
Commonly used buffering agent
HEPES is commonly used as a buffering agent.