Check Order Status
TBE Buffer, Tris-Borate-EDTA, 1X Solution, Electrophoresis, Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
TBE Buffer, Tris-Borate-EDTA, 1X Solution, Electrophoresis, Fisher BioReagents
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP2430-4
4 L
Poly CUBE
Catalog number
FBRBP2430-4
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Packaging:
Poly CUBE
Request bulk or custom format
Chemical Identifiers
CAS
77-86-1, 6381-92-6, 10043-35-3, 7732-18-5
Specifications
Chemical Name or Material
Tris-Borate-EDTA
Name Note
1X Solution
CAS
77-86-1
ChemAlert Storage Symbol
Gray
Color
Colorless
View more
Tris–Borate–EDTA (TBE) is commonly used as a buffer for nucleic acid electrophoresis.