Cesium Chloride (White Crystalline Powder/Molecular Biology), Fisher BioReagents
Cesium Chloride (White Crystalline Powder/Molecular Biology), Fisher BioReagents
Cesium Chloride (White Crystalline Powder/Molecular Biology), Fisher BioReagents
Cesium Chloride (White Crystalline Powder/Molecular Biology), Fisher BioReagents
Fisher Chemical
Cesium Chloride (White Crystalline Powder/Molecular Biology), Fisher BioReagents
Have Questions?
Catalog NumberQuantityPackaging
FBRBP210-100100 gPoly Bottle
Catalog number FBRBP210-100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Boiling Point1290°C
Chemical Name or MaterialCesium Chloride
Melting Point646°C
CAS7647-17-8
ChemAlert Storage SymbolGray
View more
This Molecular Biology-grade cesium chloride is ideal for UV analysis of nucleic acids during density-gradient centrifugation and for the isolation of plasmid DNA, phage DNA, virus particles, and RNA.