Proteinase K (Tritirachium album/Molecular Biology)
Proteinase K (<i>Tritirachium album</i>/Molecular Biology)
Actual product may vary
Fisher Chemical
Proteinase K (Tritirachium album/Molecular Biology)
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
FBRBP1700-100100 mg
FBRBP1700-50
FGCBP1700-100
FBRBP1700-500
Catalog number FBRBP1700-100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Proteinase K (Tritirachium album/Molecular Biology)
  • Not on TSCA inventory: for research and development use only; not for manufacturing or commercial purposes
Proteinase K is widely used in the isolation of DNA and RNA from cell and tissue preparations. It is a wide-spectrum protease, inactivating many enzymes, including endogenous nucleases.
Specifications
ColorWhite
For Use With (Application)Isolation of DNA and RNA from cell and tissue preparations, inactivating many Enzymes, including Endogenous Nucleases
SourceTritirachium album
Quantity100 mg
Product TypeProteinase K
FormSolid
Unit Size
Contents & Storage
-20°C