Check Order Status
Isopropyl-β-D-thiogalactopyranoside (IPTG) (Wht. Powd., Dioxane Crystalline), Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Isopropyl-β-D-thiogalactopyranoside (IPTG) (Wht. Powd., Dioxane Crystalline), Fisher BioReagents™
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP1620-10
10 g
Glass Bottle
Catalog number
FBRBP1620-10
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Isopropyl-β-D-thiogalactopyranoside
Melting Point
122°C
CAS
367-93-1
ChemAlert Storage Symbol
Gray
Color
White
View more