Check Order Status
Tris-Glycine Buffer, 10X Powder, Electrophoresis, Fisher BioReagents
Element Search
Structure Search
Tris-Glycine Buffer, 10X Powder, Electrophoresis, Fisher BioReagents
Have Questions?
Catalog Number
Quantity
FBRBP1307-1
1 Pk.
Catalog number
FBRBP1307-1
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 Pk.
Request bulk or custom format
Chemical Identifiers
CAS
77-86-1, 56-40-6
Specifications
Chemical Name or Material
Tris-Glycine
Name Note
10X Powder
CAS
77-86-1
ChemAlert Storage Symbol
Gray
Color
White
View more
Used to make a 10X concentrate solution that can be diluted for use as a transfer buffer for Western blotting and gel electrophoresis
DNase, RNase, and protease free powder that is pre-mixed for your convenience to make 1L of 10X solution
Eliminates the need to separately weigh ingredients - simply dissolve powder in pure water