Check Order Status
Vortexer Mixer Replacement Central Pad
Actual product may vary
Vortexer Mixer Replacement Central Pad
Replacement Central Pad for the Vortexer.
Have Questions?
Catalog number
FBA120328
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Vortexer Replacement Central Pad
Specifications
Product Type
Replacement Central Pad
Color
Black
Model
120328
For Use With (Equipment)
Vortexer Mixer