Surface Treated Sterile Tissue Culture Flasks, Vented Cap
Surface Treated Sterile Tissue Culture Flasks, Vented Cap
Actual product may vary
Fisherbrand
Surface Treated Sterile Tissue Culture Flasks, Vented Cap
Perfect for cell growth, cell yields aim on little or medium input volume.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Culture AreaClosure Type
FB01293550 mL25 cm²Vented
LSVFB01293550 mL25 cm²Vented
FB012937250 mL75 cm²Vented
FISFB012937250 mL75 cm²Vented
LSVFB012937250 mL75 cm²Vented
FB012939600 mL182 cm2Vented
LSVFB012939600 mL182 cm2Vented
FISFB012941850 mL300 cm2Vented
Catalog number FB012935
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
50 mL
Culture Area:
25 cm²
Closure Type:
Vented
  • Flat, free flask surface
  • Striation free to maximize usable growth area
  • Used in both open, closed systems
  • Angled neck design offers good pipet and cell scraper access
  • Upper triangular, wider base shape provides stability
  • Protruding ridge on back side of flask makes it easy for stacking
  • Special area near neck for easy mark writing
  • Both flask sides have engraved graduation
  • Sterilized by gamma irradiation
  • Non-pyrogenic
  • Vented closures
  • Vented cap filter is PVDF; pore size: 0.22μm
Specifications
Certifications/ComplianceCE
Closure ColorBlue
ColorNatural
Capacity (Metric)50 mL
For Use With (Application)Cell growth
Neck StyleAngled
No. per Case200 Flasks
No. per Pack10 Flasks
Product TypeTissue Culture Flask
Surface CoatingSurface-treated
Writing SurfaceFlat
GraduatedYes
SterilitySterile
Bottom ShapeFlat
Closure TypeVented
Culture Area25 cm²
Unit Size