Check Order Status
Assembled Screw Cap with Hole
Actual product may vary
Assembled Screw Cap with Hole
The septa and closure was designed to provide a positive physical lock to assure that preassembled components arrive intact.
Have Questions?
Catalog Number
For Use With (Equipment)
Color
ERW27020
15 mL Vial
Black
Catalog number
ERW27020
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
15 mL Vial
Color:
Black
The septa and closure was designed to provide a positive physical lock to assure that preassembled components arrive intact.
Screw caps are available for use with sealing septa.
Polypropylene caps have a maximum temperature of 135° C.
The PTFE/silicone septum can be used with most organics (until pierced) and is not recommended for chlorosilanes.
Specifications
Color
Black
Material
PTFE, Silicone, Polypropylene
No. per Pack
100
Thickness (Metric)
1.5 mm
Closure Size
18-400
For Use With (Equipment)
15 mL Vial