Barnstead™ Easypure™ Cartridges and Filters
Barnstead™ Easypure™ Cartridges and Filters
Actual product may vary
Thermo Fisher Scientific
Barnstead™ Easypure™ Cartridges and Filters
Extend the life of your Thermo Scientific™ Barnstead™ Easypure™ II or Easypure™ RF water system with replacement cartridges and filters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Description
D50231EasyPure™ II compact ultrapure water systemBarnstead™ Organic/Ion Exchange Pretreatment Cartridge, For EasyPure II Compact Ultrapure Water System
D502124Easypure™ II UV and Easypure II UV/UFCartridge pack; deionized feed
D50229EasyPure™ II water purification systemBarnstead™ Organic-free Organic Removal for Easypure II
D50233EasypureCleaning Cartridge, Easypure Ultrapure
D502125Easypure™ II UV and Easypure II UV/UFCartridge pack; RO or distilled feed
D502126Easypure™ II and Easypure UFCartridge pack; deionized feed
FL1192X1Easypure™ UF and Easypure II UV/UFUltrafilter
D50230Easypure II System, 2/3 Macroreticular Resin and 1/3 Granular, Activated Carbon, Organic/Colloidal RemovalOrganic/Colloidal removal
Catalog number D50231
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
EasyPure™ II compact ultrapure water system
Description:
Barnstead™ Organic/Ion Exchange Pretreatment Cartridge, For EasyPure II Compact Ultrapure Water System
Extend the life of your Thermo Scientific™ Barnstead™ Easypure™ II or Easypure™ RF water system with Barnstead™ Easypure™ Cartridges and Filters. Choose the cartridge pack according to your feed water and system.
D502126:
  • Includes: Organic/Colloid; two Thermo Scientific™ Ultrapure™ Ion Exchange
  • Type 1 filter
D502124:
  • Includes: Organic/Colloid, Ultrapure Ion Exchange, Organic Free Organic Removal
  • Ultralow filter with Organic Free Cartridge
D502127:
  • Includes: Organic/Ion Exchange, two Ultrapure Ion Exchange
  • Type 1 filter
D502125:
  • Includes: 09-034-18 Organic/Ion Exchange, 09-050-46 Ultrapure Ion Exchange, 09-034-16 Organic Free Organic Removal
  • Ultralow filter with Organic Free Cartridge
Specifications
DescriptionBarnstead™ Organic/Ion Exchange Pretreatment Cartridge, For EasyPure II Compact Ultrapure Water System
Feedwater SourcePretreated
Resin TypeActivated Carbon and Mixed Bed Ion Exchange Resin
For Use With (Application)Organic/ion exchange cartridge
For Use With (Equipment)EasyPure™ II compact ultrapure water system
TypeCartridge and Filter
Unit Size