Search Thermo Fisher Scientific
Catalog Number | For Use With (Equipment) | Resin Type | Capacity | For Use With (Application) |
---|---|---|---|---|
D0836 | B-Pure™ | Macroreticular, Activated Carbon | 2000 gal. | Pretreatment Macropure |
D0809 | Bantam™ Deionizer, B-Pure™ | Mixed Bed | 875 grains | |
D0811 | B-Pure™ | Porous Strong Ion Exchange | 30 g | Oxygen Removal |
D0813 | Bantam™ Deionizer, B-Pure™ | Activated Carbon | 5000 gal. | Organic Removal |
D0832 | Bantam™ Deionizer, B-Pure™ | Mixed Bed, Activated Carbon | 785 grains/1000 gal. | Ultrapure and Organic Removal |
D0835 | B-Pure™ | Macroreticular, Activated Carbon | 5000 gal. | Pretreatment |
FIS09-034-5C | B-Pure™ | Macroreticular, Activated Carbon | 2000 gal. | Pretreatment Macropure |
D63112 | B-Pure™ | Extruded Carbon | 5000 gal. | Organic Removal |
D8811 | B-Pure™ | Ultrapure, Oxygen and Organic Removal | 365 grains/2000 gal./12 g | Ultrapure, Oxygen and Organic Removal |
D0803 | Thermo Scientific™ Barnstead™ Bantam™ B-Pure™ or Nanopure™ Water Purification Systems as Indicated | Two Bed Ion Exchange | 1760 grains | Cartridge for high capacity; two-bed ion exchange; full size |