1,3-Bis(4-piperidinyl)propane, 97+%, Thermo Scientific Chemicals
1,3-Bis(4-piperidinyl)propane, 97+%, Thermo Scientific Chemicals
1,3-Bis(4-piperidinyl)propane, 97+%, Thermo Scientific Chemicals
1,3-Bis(4-piperidinyl)propane, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3-Bis(4-piperidinyl)propane, 97+%, Thermo Scientific Chemicals
CAS: 16898-52-5 | C13H26N2 | 210.365 g/mol
Have Questions?
Catalog NumberQuantity
ALFL20442.1425 g
Catalog number ALFL20442.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material1,3-Bis(4-piperidinyl)propane
CAS16898-52-5
Health Hazard 1H302-H314-H335
Health Hazard 2GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c