Spermine, 97%, Thermo Scientific Chemicals
Spermine, 97%, Thermo Scientific Chemicals
Spermine, 97%, Thermo Scientific Chemicals
Spermine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Spermine, 97%, Thermo Scientific Chemicals
CAS: 71-44-3 | C10H26N4 | 202.35 g/mol
Have Questions?
Catalog NumberQuantity
ALFL19562.031 g
Catalog number ALFL19562.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS18851-33-7
IUPAC Name1,10-phenanthroline hydrate hydrochloride
Molecular FormulaC12H11ClN2O
InChI KeyNDLHUHRGAIHALB-UHFFFAOYSA-N
SMILESO.Cl.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=96.0 %
Sulfated ash=<0.2 %
Appearance (Color)White to light yellow to light purple
Infrared spectrumConforms
Appearance (Form)Powder or crystals
View more
Binds to the polyamine modulatory site of NMDA Spermine is essential for both normal and neoplastic tissue growth. It is involved in the modulation of calcium-dependent immune processes. It plays an important role in cellular proliferation and differentiation as well as inhibits neuronal nitric oxide synthase (nNOS).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Binds to the polyamine modulatory site of NMDA Spermine is essential for both normal and neoplastic tissue growth. It is involved in the modulation of calcium-dependent immune processes. It plays an important role in cellular proliferation and differentiation as well as inhibits neuronal nitric oxide synthase (nNOS).
Solubility
Soluble in chloroform, methanol, water and lower alcohols. Insoluble in ether, benzene and petroleum ether.
Notes
Air sensitive and hygroscopic. Store in a cool place. Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
RUO – Research Use Only
General References:
  1. Bailey, D.; Kirby, B. P.; Atkinson, J.; Fixon-Owoo, S.; Henman, M. C.; Shaw, G. G.; Doyle, K. M. Hydroxycinnamic acid amide derivatives of polyamines reverse spermine-induced CNS excitation. Pharmacol. Biochem. Behav. 2015, 133, 57-64.
  2. Mo, H.; Wang, X.; Zhang, Y.; Zhang, G.; Zhang, J.; Ma, Z. Cotton polyamine oxidase is required for spermine and camalexin signalling in the defence response to Verticillium dahliae. Plant J. 2015, 83 (6), 962-975.