1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether, 97%, Thermo Scientific Chemicals
1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether, 97%, Thermo Scientific Chemicals
1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether, 97%, Thermo Scientific Chemicals
1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether, 97%, Thermo Scientific Chemicals
CAS: 26675-46-7 | C3H2ClF5O | 184.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFL17315.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether
CAS26675-46-7
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335-H336
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether is a useful solvent for dispersing fluorinated materials.
For Research and Development use only. Not to be used in clinical studies