1,5-Pentamethylene-1H-tetrazole, 98%, Thermo Scientific Chemicals
1,5-Pentamethylene-1H-tetrazole, 98%, Thermo Scientific Chemicals
1,5-Pentamethylene-1H-tetrazole, 98%, Thermo Scientific Chemicals
1,5-Pentamethylene-1H-tetrazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,5-Pentamethylene-1H-tetrazole, 98%, Thermo Scientific Chemicals
CAS: 54-95-5 | C6H10N4 | 138.174 g/mol
Have Questions?
Catalog NumberQuantity
ALFL16185.065 g
Catalog number ALFL16185.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7727-54-0
IUPAC Namediammonium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular FormulaH8N2O8S2
InChI KeyROOXNKNUYICQNP-UHFFFAOYSA-N
SMILES[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Titration with KMnO4>=98.0 %
Heavy metals (as Pb)=<0.005 %
Residue after ignition=<0.05 %