Glycolic acid, ca 66-70% aq. soln., Thermo Scientific Chemicals
Glycolic acid, ca 66-70% aq. soln., Thermo Scientific Chemicals
Glycolic acid, ca 66-70% aq. soln., Thermo Scientific Chemicals
Glycolic acid, ca 66-70% aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycolic acid, ca 66-70% aq. soln., Thermo Scientific Chemicals
CAS: 79-14-1 | C2H4O3 | 76.051 g/mol
Have Questions?
Catalog NumberQuantity
ALFL14980.AP500 mL
Catalog number ALFL14980.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialGlycolic acid
CAS79-14-1
Boiling Point112°C
Health Hazard 1H314-H332-H335
Health Hazard 2GHS H Statement
H314-H302-H332
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
View more
Glycolic acid is used to evaluate the efficacy of glycolic peels treatment for all types of acne. It is also used in the fine synthesis of medicine and as a raw material of cosmetics and organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycolic acid is used to evaluate the efficacy of glycolic peels treatment for all types of acne. It is also used in the fine synthesis of medicine and as a raw material of cosmetics and organic synthesis.
Solubility
Soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Incompatible with bases and oxidizing agents.
RUO – Research Use Only
General References:
  1. Alicia Garcia Bs.; James E. Fulton. The Combination of Glycolic Acid and Hydroquinone or Kojic Acid for the Treatment of Melasma and Related Conditions. Dermatologic Surgery. 1996, 22 (5), 443-447.
  2. Tolbert, N. E.; Zill, L. P. Excretion of glycolic acid by algae during photosynthesis. Journal of Biological Chemistry. 1956, 222 (2), 895-906.