Ethyl 4-hydroxy-3-methoxycinnamate, 98%, Thermo Scientific Chemicals
Ethyl 4-hydroxy-3-methoxycinnamate, 98%, Thermo Scientific Chemicals
Ethyl 4-hydroxy-3-methoxycinnamate, 98%, Thermo Scientific Chemicals
Ethyl 4-hydroxy-3-methoxycinnamate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl 4-hydroxy-3-methoxycinnamate, 98%, Thermo Scientific Chemicals
CAS: 4046-02-0 | C12H14O4 | 222.24 g/mol
Have Questions?
Catalog NumberQuantity
ALFL14960.031 g
Catalog number ALFL14960.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Density(20/20°C) 0.784 to 0.787
Appearance (Form)Clear liquid
Color scale=<10 APHA
GC>=99.5 %
Water=<0.2 % (K.F.)