Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride, 97%, Thermo Scientific Chemicals
Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride, 97%, Thermo Scientific Chemicals
Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride, 97%, Thermo Scientific Chemicals
Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride, 97%, Thermo Scientific Chemicals
CAS: 81-30-1 | C14H4O6 | 268.18 g/mol
Have Questions?
Catalog NumberQuantity
ALFL14365.065 g
Catalog number ALFL14365.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS6484-52-2
IUPAC Nameammonium nitrate
Molecular FormulaH4N2O3
InChI KeyDVARTQFDIMZBAA-UHFFFAOYSA-O
SMILES[NH4+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Adhering crystals
pH4.5 to 6 (5 % at 20°C)
Iron (Fe)=<2 ppm
Heavy metals (as Pb)=<5 ppm
View more
Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride is used as a reagent in the preparation of novel fluorescent markers for hypoxic cells of naphthalimides with two heterocyclic side chains for bioreductive binding.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride is used as a reagent in the preparation of novel fluorescent markers for hypoxic cells of naphthalimides with two heterocyclic side chains for bioreductive binding.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Store under inert gas. Incompatible with acids, bases and oxidizing agents.
RUO – Research Use Only
Yanqing Xu.; Daqiang Yuan.; Benlai Wu.; Feilong Jiang.; Youfu Zhou.; Maochun Hong. Two photoluminescent coordination polymers based on naphthalene-1,4,5,8-tetracarboxylic acid 4,5-anhydride. Inorganic Chemistry Communications. 2005, 8 (7), 651-655.