2-Aminoanthracene, 94%, Thermo Scientific Chemicals
2-Aminoanthracene, 94%, Thermo Scientific Chemicals
2-Aminoanthracene, 94%, Thermo Scientific Chemicals
2-Aminoanthracene, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Aminoanthracene, 94%, Thermo Scientific Chemicals
CAS: 613-13-8 | C14H11N | 193.249 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFL14269.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Aminoanthracene
CAS613-13-8
Health Hazard 1H341
Health Hazard 2GHS H Statement
H341
Suspected of causing genetic defects.
Health Hazard 3P201-P202-P281-P308+P313-P501c