3',4',5,7-Tetrahydroxyflavone, 97%, Thermo Scientific Chemicals
3',4',5,7-Tetrahydroxyflavone, 97%, Thermo Scientific Chemicals
3',4',5,7-Tetrahydroxyflavone, 97%, Thermo Scientific Chemicals
3',4',5,7-Tetrahydroxyflavone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3',4',5,7-Tetrahydroxyflavone, 97%, Thermo Scientific Chemicals
CAS: 491-70-3 | C15H10O6 | 286.239 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
100 mg
Catalog number ALFL14186.ME
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Request bulk or custom format
Chemical Identifiers
CAS1591-31-7
IUPAC Name4-iodo-1,1'-biphenyl
Molecular FormulaC12H9I
InChI KeyNXYICUMSYKIABQ-UHFFFAOYSA-N
SMILESIC1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay (GC)≥96.0%
Melting Point (clear melt)108.0-118.0?C
3',4',5,7-Tetrahydroxyflavone is used as an apoptosis inducer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3′,4′,5,7-Tetrahydroxyflavone is used as an apoptosis inducer.
Solubility
Soluble in aqueous alkaline solutions (1.4 mg/ml), ethanol (∼5 mg/ml), dimethyl sulfoxide (7 mg/ml), 1eq. Sodium hydroxide (5 mM), dimethylformamide (∼20 mg/ml), water (1 mg/ml) at 25°C and methanol.
Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C.
RUO – Research Use Only
General References:
  1. NK Peters.; JW Frost.; SR Long. A plant flavone, luteolin, induces expression of Rhizobium meliloti nodulation genes. Science. 1986, 233 (4767), 977-980.
  2. Seelinger G.; Merfort I.; Schempp CM. Anti-oxidant, anti-inflammatory and anti-allergic activities of luteolin. Planta Medica. 2007, 208 (15), 1694-1706.
  3. Anti-oxidant and radical scavenger in biological systems: Methods Enzymol., 234, 420 (1994).