(S)-(+)-2-Butanol, 98+%, Thermo Scientific Chemicals
(S)-(+)-2-Butanol, 98+%, Thermo Scientific Chemicals
(S)-(+)-2-Butanol, 98+%, Thermo Scientific Chemicals
(S)-(+)-2-Butanol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(S)-(+)-2-Butanol, 98+%, Thermo Scientific Chemicals
CAS: 4221-99-2 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFL14164.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS69-89-6
IUPAC Name2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular FormulaC5H4N4O2
InChI KeyLRFVTYWOQMYALW-UHFFFAOYSA-N
SMILESO=C1NC2=C(NC=N2)C(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay from Supplier's CofA≥98.5% (HPLC)
FormPowder
Infrared spectrumConforms
For synthesis of optically active products