Silica gel 60, 0.032-0.063mm (230-450 mesh), Thermo Scientific Chemicals
Silica gel 60, 0.032-0.063mm (230-450 mesh), Thermo Scientific Chemicals
Silica gel 60, 0.032-0.063mm (230-450 mesh), Thermo Scientific Chemicals
Silica gel 60, 0.032-0.063mm (230-450 mesh), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silica gel 60, 0.032-0.063mm (230-450 mesh), Thermo Scientific Chemicals
CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1000 g
2500 g
Catalog number ALFL14003.36
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
Assay from Supplier's CofA≥99.999% (metals basis)
FormSlug, 3.175mm (0.125in) diameter.
Length3.175mm (0.125in)
Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.
Solubility
Soluble in hydrofluoric acid. Insoluble in water.
Notes
Keep the container tightly sealed. Store in cool, dry conditions in a well sealed container. Incompatible with hydrogen fluoride.
RUO – Research Use Only
General References:
  1. Suitable for conventional and flash column chromatography.
  2. Marchenko, M.; Thomson, A.; Ellis, T. N.; Knuckley, B.; Causey, C. P. Development of a clickable activity-based protein profiling (ABPP) probe for agmatine deiminases. Bioorg. Med. Chem. 2015, 23 (9), 2159-2167.
  3. Thomson, A.; O’Connor, S.; Knuckley, B.; Causey, C. P. Design, synthesis, and in vitro evaluation of an activity-based protein profiling (ABPP) probe targeting agmatine deiminases. Bioorg. Med. Chem. 2014, 22 (17), 4602-4608.