N-tert-Butyl-2-benzothiazolesulfenamide, 97%, Thermo Scientific Chemicals
N-tert-Butyl-2-benzothiazolesulfenamide, 97%, Thermo Scientific Chemicals
N-tert-Butyl-2-benzothiazolesulfenamide, 97%, Thermo Scientific Chemicals
N-tert-Butyl-2-benzothiazolesulfenamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-tert-Butyl-2-benzothiazolesulfenamide, 97%, Thermo Scientific Chemicals
CAS: 95-31-8 | C11H14N2S2 | 238.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALFL13356.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS452-58-4
IUPAC Namepyridine-2,3-diamine
Molecular FormulaC5H7N3
InChI KeyZZYXNRREDYWPLN-UHFFFAOYSA-N
SMILESNC1=CC=CN=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to dark brown
AppearanceMay darken on storage
Titration with HClO497.5 to 102.5 %
GC>=97.5 %
Appearance (Form)Crystalline powder or needles