Tetra-n-butylammonium fluoride trihydrate, 98%, Thermo Scientific Chemicals
Tetra-n-butylammonium fluoride trihydrate, 98%, Thermo Scientific Chemicals
Tetra-n-butylammonium fluoride trihydrate, 98%, Thermo Scientific Chemicals
Tetra-n-butylammonium fluoride trihydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetra-n-butylammonium fluoride trihydrate, 98%, Thermo Scientific Chemicals
CAS: 87749-50-6 | C16H42FNO3 | 315.514 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number ALFL13303.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS99-63-8
IUPAC Namebenzene-1,3-dicarbonyl dichloride
Molecular FormulaC8H4Cl2O2
InChI KeyFDQSRULYDNDXQB-UHFFFAOYSA-N
SMILESClC(=O)C1=CC(=CC=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline mass
Infrared spectrumConforms
Melting point43°C to 45°C
Titration RCOCl>=97.5 %
Tetra-n-butylammonium fluoride trihydrate is used as a phase transfer catalyst, as a mild base and as a source of fluoride ion in organic solvents. It is also used as a deprotecting agent to remove silyl ether protecting groups. It acts as a reactant for the preparation of deprotecting agents, in preparation of cellulose derivatives, lipophilic peptides for DNA transfections and Dehydrobromination reactions. It plays an important role in the convertion of O-silylated enolates in to carbonyls in the presence of dimethyl sulfoxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetra-n-butylammonium fluoride trihydrate is used as a phase transfer catalyst, as a mild base and as a source of fluoride ion in organic solvents. It is also used as a deprotecting agent to remove silyl ether protecting groups. It acts as a reactant for the preparation of deprotecting agents, in preparation of cellulose derivatives, lipophilic peptides for DNA transfections and Dehydrobromination reactions. It plays an important role in the convertion of O-silylated enolates in to carbonyls in the presence of dimethyl sulfoxide.
Solubility
Soluble in water, terahydrofuran, dimethyl sulfoxide and acetonitrile.
Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Huang, S. H.; Liu, C. H.; Yang, C. M. Efficient ligand-free Hiyama cross-coupling reaction catalyzed by functionalized SBA-15-supported Pd nanoparticles. Green Chem. 2014, 16 (5), 2706-2712.
  2. Liang, X.; Kozlovskaya, V.; Cox, C. P.; Wang, Y.; Saeed, M.; Kharlampieva, E. Synthesis and self-assembly of thermosensitive double-hydrophilic poly(N-vinylcaprolactam)-b-poly(N-vinyl-2-pyrrolidone) diblock copolymers. J. Polym. Sci., Part A: Polym. Chem. 2014, 52 (19), 2725-2737.