4,6-Diamino-2-hydroxy-1,3,5-triazine, tech. 90%, Thermo Scientific Chemicals
4,6-Diamino-2-hydroxy-1,3,5-triazine, tech. 90%, Thermo Scientific Chemicals
4,6-Diamino-2-hydroxy-1,3,5-triazine, tech. 90%, Thermo Scientific Chemicals
4,6-Diamino-2-hydroxy-1,3,5-triazine, tech. 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4,6-Diamino-2-hydroxy-1,3,5-triazine, tech. 90%, Thermo Scientific Chemicals
CAS: 645-92-1 | C3H5N5O | 127.107 g/mol
Have Questions?
Catalog NumberQuantity
ALFL13064.031 g
Catalog number ALFL13064.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS6363-53-7
IUPAC Name2,3,5,6-tetrahydroxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
Molecular FormulaC12H24O12
InChI KeyHBDJFVFTHLOSDW-UHFFFAOYNA-N
SMILESO.OCC(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Additional inforemainder mainly glucose and maltotriose
Sulfated ash=<0.1 %
Specific optical rotation+135° to +139° (20°C, 589 nm) (c=4, H2O, NH3)
Specific optical rotationon dry substance