Domiphen bromide, 97%, Thermo Scientific Chemicals
Domiphen bromide, 97%, Thermo Scientific Chemicals
Domiphen bromide, 97%, Thermo Scientific Chemicals
Domiphen bromide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Domiphen bromide, 97%, Thermo Scientific Chemicals
CAS: 538-71-6 | C22H40BrNO | 414.472 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFL12930.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS194491-31-1
IUPAC Nametetrasodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Molecular FormulaC10H12N2Na4O8
InChI KeyUEUXEKPTXMALOB-UHFFFAOYSA-J
SMILES[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms (UK Sourced Material)
CommentMaterial Sourced in UK and US
Water Content (Karl Fischer Titration)8-17% (ca. 2-4 waters) (UK Sourced Material)
FormPowder or crystalline powder
Assay from Supplier's CofA≥97.5% (US Sourced Material)
View more
Domiphen bromide is employed as a phase transfer catalyst in synthetic chemistry. It is used as a quaternary ammonium cation with cationic surfactant properties. Further, it is used as an antiseptic and an active pharmaceutical ingredient.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Domiphen bromide is employed as a phase transfer catalyst in synthetic chemistry. It is used as a quaternary ammonium cation with cationic surfactant properties. Further, it is used as an antiseptic and an active pharmaceutical ingredient.
Solubility
Soluble in ethanol, acetone, ethyl acetate, chloroform and water. Slightly soluble in benzene.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Mester, P.; Gundolf, T.; Kalb, R.; Wagner, M.; Rossmanith, P. Molecular mechanisms mediating tolerance to ionic liquids in Listeria monocytogenes. Sep. Purif. Technol. 2015, 155, 32-37.
  2. Nestola, P.; Peixoto, C.; Silva, R. R. J. S.; Alves, P. M.; Mota, J. P. B.; Carrondo, M. J. T. Improved virus purification processes for vaccines and gene therapy. Biotechnol. Bioeng. 2015, 112 (5), 843-857.