4-Nitrophenyl octanoate, 96%, Thermo Scientific Chemicals
4-Nitrophenyl octanoate, 96%, Thermo Scientific Chemicals
4-Nitrophenyl octanoate, 96%, Thermo Scientific Chemicals
4-Nitrophenyl octanoate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrophenyl octanoate, 96%, Thermo Scientific Chemicals
CAS: 1956-10-1 | C14H19NO4 | 265.31 g/mol
Have Questions?
Catalog NumberQuantity
ALFL12022.065 g
Catalog number ALFL12022.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Nitrophenyl octanoate
CAS1956-10-1
Recommended StorageAmbient temperatures
Density1.082
TSCANo