Dibenzyl disulfide, 98+%, Thermo Scientific Chemicals
Dibenzyl disulfide, 98+%, Thermo Scientific Chemicals
Dibenzyl disulfide, 98+%, Thermo Scientific Chemicals
Dibenzyl disulfide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dibenzyl disulfide, 98+%, Thermo Scientific Chemicals
CAS: 150-60-7 | C14H14S2 | 246.386 g/mol
Have Questions?
Catalog NumberQuantity
ALFL11850.22100 g
Catalog number ALFL11850.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS1460-57-7
IUPAC Name(1R,2R)-cyclohexane-1,2-diol
Molecular FormulaC6H12O2
InChI KeyPFURGBBHAOXLIO-PHDIDXHHSA-N
SMILESO[C@@H]1CCCC[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)99.0-106.0?C
Appearance (Color)White to cream
Assay (GC)≥97.5%