3-Methyl-1-pentene, 98%, Thermo Scientific Chemicals
3-Methyl-1-pentene, 98%, Thermo Scientific Chemicals
3-Methyl-1-pentene, 98%, Thermo Scientific Chemicals
3-Methyl-1-pentene, 98%, Thermo Scientific Chemicals
3-Methyl-1-pentene, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFL11525.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Methyl-1-pentene
CAS760-20-3
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Health Hazard 3GHS P Statement
P210-P243-P301+P330+P331-P315
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Take precautionary measures against static discharge.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Get immediate medical advice/attention.