trans-1,2-Cyclohexanedicarboxylic acid, 98%, Thermo Scientific Chemicals
trans-1,2-Cyclohexanedicarboxylic acid, 98%, Thermo Scientific Chemicals
trans-1,2-Cyclohexanedicarboxylic acid, 98%, Thermo Scientific Chemicals
trans-1,2-Cyclohexanedicarboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-1,2-Cyclohexanedicarboxylic acid, 98%, Thermo Scientific Chemicals
CAS: 2305-32-0 | C8H12O4 | 172.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFL11288.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS41532-84-7
IUPAC Name1,1,2-trimethyl-1H-benzo[e]indole
Molecular FormulaC15H15N
InChI KeyWJZSZXCWMATYFX-UHFFFAOYSA-N
SMILESCC1=NC2=CC=C3C=CC=CC3=C2C1(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Infrared spectrumConforms
HPLC>=95 %
Appearance (Form)Crystalline powder
Melting point111°C to 117°C