Phloretin, 98%, Thermo Scientific Chemicals
Phloretin, 98%, Thermo Scientific Chemicals
Phloretin, 98%, Thermo Scientific Chemicals
Phloretin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phloretin, 98%, Thermo Scientific Chemicals
CAS: 60-82-2 | C15H14O5 | 274.272 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
25 mg
Catalog number ALFL10991.MC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS1129-26-6
IUPAC Name4-methoxybenzene-1-sulfonamide
Molecular FormulaC7H9NO3S
InChI KeyMSFQEZBRFPAFEX-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)S(N)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
HPLC>=94.0 %
Appearance (Form)Crystalline powder
Melting point109°C to 113°C
Topical antioxidant