Vitamin K{1}, Thermo Scientific Chemicals
Vitamin K{1}, Thermo Scientific Chemicals
Vitamin K{1}, Thermo Scientific Chemicals
Vitamin K{1}, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Vitamin K{1}, Thermo Scientific Chemicals
Important dietary source found in green plants, algae and photosynthetic bacteria. It is used as a vitamin (prothrombogenic) and veterinary antidote for dicoumarol poisoning. It is an essential vitamin for the synthesis of coagulation factors II, VII, IX, and X in the liver.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFL10575.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS868-14-4
IUPAC Namepotassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
Molecular FormulaC4H5KO6
InChI KeyKYKNRZGSIGMXFH-ZVGUSBNCSA-M
SMILES[K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Identification (FTIR)Conforms
Optical Rotation32.3 ± 0.4? (C=10 in 1N NaOH)
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Vitamin K{1} is an important dietary source found in green plants, algae and photosynthetic bacteria. It is used as a vitamin (prothrombogenic) and veterinary antidote for dicoumarol poisoning. It is an essential vitamin for the synthesis of coagulation factors II, VII, IX, and X in the liver.