1-Hexanethiol, 96%, Thermo Scientific Chemicals
1-Hexanethiol, 96%, Thermo Scientific Chemicals
1-Hexanethiol, 96%, Thermo Scientific Chemicals
1-Hexanethiol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Hexanethiol, 96%, Thermo Scientific Chemicals
CAS: 111-31-9 | C6H14S | 118.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Catalog number ALFL09659.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS24696-26-2
IUPAC NameN-(1,3-dihydroxyoctadec-4-en-2-yl)hexadecanamide
Molecular FormulaC34H67NO3
InChI KeyYDNKGFDKKRUKPY-UHFFFAOYNA-N
SMILESCCCCCCCCCCCCCCCC(=O)NC(CO)C(O)C=CCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Enantiomeric excess>=99.0 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder
HPLC>=97.50 %
It is used to tune the electrical characteristics in networked carbon nanotube field-effect transistors.1 Also used as a monolayer on copper surfaces to retard surface oxidation during exposure to the environment which can then be desorbed effectively with an annealing step in inert N2 to provide a clean Cu surface.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used to tune the electrical characteristics in networked carbon nanotube field-effect transistors.1 Also used as a monolayer on copper surfaces to retard surface oxidation during exposure to the environment which can then be desorbed effectively with an annealing step in inert N2 to provide a clean Cu surface.
Solubility
Difficult to mix in water.
Notes
Air sensitive. Store in cool dry condition. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Edith Chow.; Thomas R. Gengenbach.; Lech Wieczorek.; Burkhard Raguse. Detection of organics in aqueous solution using gold nanoparticles modified with mixed monolayers of 1-hexanethiol and 4-mercaptophenol.Sens. Actuators, B. 2010, 143 (2), 704-711 .
  2. Gary K. Rowe.; Stephen E. Creager. Chain Length and Solvent Effects on Competitive Self-Assembly of Ferrocenylhexanethiol and 1-Alkanethiols onto Gold. Langmuir. 1994, 10 (4),1186-1192 .