1-Hexanethiol, 96%, Thermo Scientific Chemicals
1-Hexanethiol, 96%, Thermo Scientific Chemicals
1-Hexanethiol, 96%, Thermo Scientific Chemicals
1-Hexanethiol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Hexanethiol, 96%, Thermo Scientific Chemicals
CAS: 111-31-9 | C6H14S | 118.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number ALFL09659.AC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Request bulk or custom format
Specifications
Chemical Name or Material1-Hexanethiol
CAS111-31-9
Health Hazard 1H226-H302-H331
Health Hazard 2GHS H Statement
H301-H331-H226
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapor.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P311-P330-P370+P378q-P501c
View more
It is used to tune the electrical characteristics in networked carbon nanotube field-effect transistors.1 Also used as a monolayer on copper surfaces to retard surface oxidation during exposure to the environment which can then be desorbed effectively with an annealing step in inert N2 to provide a clean Cu surface.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used to tune the electrical characteristics in networked carbon nanotube field-effect transistors.1 Also used as a monolayer on copper surfaces to retard surface oxidation during exposure to the environment which can then be desorbed effectively with an annealing step in inert N2 to provide a clean Cu surface.
Solubility
Difficult to mix in water.
Notes
Air sensitive. Store in cool dry condition. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Edith Chow.; Thomas R. Gengenbach.; Lech Wieczorek.; Burkhard Raguse. Detection of organics in aqueous solution using gold nanoparticles modified with mixed monolayers of 1-hexanethiol and 4-mercaptophenol.Sens. Actuators, B. 2010, 143 (2), 704-711 .
  2. Gary K. Rowe.; Stephen E. Creager. Chain Length and Solvent Effects on Competitive Self-Assembly of Ferrocenylhexanethiol and 1-Alkanethiols onto Gold. Langmuir. 1994, 10 (4),1186-1192 .