Methyl 3,4-dihydroxybenzoate, 97%, Thermo Scientific Chemicals
Methyl 3,4-dihydroxybenzoate, 97%, Thermo Scientific Chemicals
Methyl 3,4-dihydroxybenzoate, 97%, Thermo Scientific Chemicals
Methyl 3,4-dihydroxybenzoate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl 3,4-dihydroxybenzoate, 97%, Thermo Scientific Chemicals
CAS: 2150-43-8 | C8H8O4 | 168.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number ALFL09552.04
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 g
Request bulk or custom format
Chemical Identifiers
CAS1484-85-1
IUPAC Namehydrogen 2-(2H-1,3-benzodioxol-5-yl)ethan-1-amine chloride
Molecular FormulaC9H12ClNO2
InChI KeyNDYXFQODWGEGNU-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCCC1=CC=C2OCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow
Assay from Supplier's CofA≥94.0%