Fluorescein isothiocyanate, isomer 1, 95%
Fluorescein isothiocyanate, isomer 1, 95%
Fluorescein isothiocyanate, isomer 1, 95%
Fluorescein isothiocyanate, isomer 1, 95%
Thermo Scientific Chemicals
Fluorescein isothiocyanate, isomer 1, 95%
CAS: 3326-32-7 | C21H11NO5S | 389.381 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
250 mg
Catalog number ALFL09319.MF
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialFluorescein isothiocyanate
Name Noteisomer 1
CAS3326-32-7
Health Hazard 1H302-H315-H317-H319-H334-H335
Health Hazard 2GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
View more
Fluorescein isothiocyanate is used as fluorescent labeling reagent for proteins and for rapid identification of pathogens in fluorescent antibody technique. It is also used as a reagent for microsequencing of proteins and peptides (HPLC). On celite it is used for the labeling of fibrinogen.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Fluorescein isothiocyanate is used as fluorescent labeling reagent for proteins and for rapid identification of pathogens in fluorescent antibody technique. It is also used as a reagent for microsequencing of proteins and peptides (HPLC). On celite it is used for the labeling of fibrinogen.
Solubility
Slightly soluble in DMSO, ethanol, methanol DMF and acetone
Notes
Incompatible with strong oxidizing agents, strong bases and amines.
RUO – Research Use Only
General References:
  1. Reagent for fluorescent labelling of proteins: Anal. Biochem., 20, 178 (1968).
  2. Guilini, C.; Baehr, C.; Schaeffer, E.; Gizzi, P.; Rufi, F.; Haiech, J.; Weiss, E.; Bonnet, D.; Galzi, J. New Fluorescein Precursors for Live Bacteria Detection. Anal. Chem. 2015, 87 (17), 8858-8866.
  3. Chen, Y.; Yang, C.; Li, T.; Zhang, M.; Liu, Y.; Gauthier, M. A.; Zhao, Y.; Wu, C. The Interplay of Disulfide Bonds, α-Helicity, and Hydrophobic Interactions Leads to Ultrahigh Proteolytic Stability of Peptides. Biomacromolecules 2015, 16 (8), 2347-2355.