3-Chlorobenzenesulfonamide, 98%, Thermo Scientific Chemicals
3-Chlorobenzenesulfonamide, 98%, Thermo Scientific Chemicals
3-Chlorobenzenesulfonamide, 98%, Thermo Scientific Chemicals
3-Chlorobenzenesulfonamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Chlorobenzenesulfonamide, 98%, Thermo Scientific Chemicals
CAS: 17260-71-8 | C6H6ClNO2S | 191.63 g/mol
Have Questions?
Catalog NumberQuantity
ALFL08349.031 g
Catalog number ALFL08349.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Chlorobenzenesulfonamide
CAS17260-71-8
Melting Point144°C to 148°C
Recommended StorageAmbient temperatures
TSCANo
View more
It can react with 1-chloro-4-isocyanato-benzene to prepare C13H10Cl2N2O3S in the presence of 1 N aq. NaOH and solvent acetone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can react with 1-chloro-4-isocyanato-benzene to prepare C13H10Cl2N2O3S in the presence of 1 N aq. NaOH and solvent acetone.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only