(+/-)-Styrene oxide, 98+%, Thermo Scientific Chemicals
(+/-)-Styrene oxide, 98+%, Thermo Scientific Chemicals
(+/-)-Styrene oxide, 98+%, Thermo Scientific Chemicals
(+/-)-Styrene oxide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(+/-)-Styrene oxide, 98+%, Thermo Scientific Chemicals
CAS: 96-09-3 | C8H8O | 120.151 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFL07821.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material(+/-)-Styrene oxide
CAS96-09-3
Health Hazard 1H227-H312-H315-H317-H319-H331-H340-H350
Health Hazard 2GHS H Statement
H350-H302-H312-H319-H227
May cause cancer.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Combustible liquid.
Health Hazard 3P201-P202-P210-P235-P261-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P333+P313-P362-P370+P378q-P501c
View more
Styrene oxide is used as an intermediate in medicine and perfume.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Styrene oxide is used as an intermediate in medicine and perfume.
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Aryl substituted epoxides undergo rearrangement in the presence of NaBH4 in benzene to give arylacetaldehydes: Gazz. Chim. Ital., 121, 579 (1991).
  2. In the presence of Pd(OAc)2 and Bu3P, a tandem isomerization-aldol condensation occurs: J. Org. Chem., 61, 7565 (1996):
  3. In the presence of excess of a second aldehyde, crossed aldol reactions occur in moderate yield.