1,3,5-Benzenetricarbonyl chloride, 98+%, Thermo Scientific Chemicals
1,3,5-Benzenetricarbonyl chloride, 98+%, Thermo Scientific Chemicals
1,3,5-Benzenetricarbonyl chloride, 98+%, Thermo Scientific Chemicals
1,3,5-Benzenetricarbonyl chloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3,5-Benzenetricarbonyl chloride, 98+%, Thermo Scientific Chemicals
CAS: 4422-95-1 | C9H3Cl3O3 | 265.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFL07255.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS96-09-3
IUPAC Name2-phenyloxirane
Molecular FormulaC8H8O
InChI KeyAWMVMTVKBNGEAK-UHFFFAOYNA-N
SMILESC1OC1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
Refractive Index1.5330-1.5370 @20°C
1,3,5-Benzenetricarbonyl chloride was used in the synthesis of chiral azoaromatic dendrimeric systems. It was used to study the structure of activated composite membranes containing organophosphorus extractants as carriers. It was the starting material for two tritopic amides derived from 3- and 4-methylaminopyridine which self-assembled into nanoballs on treatment with palladium(II) nitrate in DMSO.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3,5-Benzenetricarbonyl chloride was used in the synthesis of chiral azoaromatic dendrimeric systems. It was used to study the structure of activated composite membranes containing organophosphorus extractants as carriers. It was the starting material for two tritopic amides derived from 3- and 4-methylaminopyridine which self-assembled into nanoballs on treatment with palladium(II) nitrate in DMSO.
Solubility
Soluble in chloroform(soluble10%, clear to very slightly hazy, colorless to faintly yellow) and toluene. Solubility in water: Reacts.
Notes
Moisture Sensitive. Store away from strong bases and oxidizing agents. Incompatible with water, bases and oxidizing agents. Store at 4°C.
RUO – Research Use Only
General References:
  1. Luigi Angiolini; Tiziana Benelli; Loris Giorgini. Synthesis and chiroptical properties of chiral azoaromatic dendrimers with a C3-symmetrical core.Chirality. 2010, 22 (1), 99-109.
  2. Kranthikumar Tungala; P. Adhikary; S. Krishnamoorthi. Trimerization of β-cyclodextrin through the click reaction.Carbohydrate Polymers.2013, 95 (1), 295-298.