1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific Chemicals
1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific Chemicals
1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific Chemicals
1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific Chemicals
CAS: 95-93-2 | C10H14 | 134.222 g/mol
Have Questions?
Catalog NumberQuantity
ALFL04493.22100 g
Catalog number ALFL04493.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material1,2,4,5-Tetramethylbenzene
CAS95-93-2
Health Hazard 1H228
Health Hazard 2GHS H Statement
H228
Flammable solid.
Health Hazard 3P210-P240-P241-P280-P370+P378q
View more
1,2,4,5-Tetramethylbenzene is the precursor to pyromellitic dianhydride, which is used for manufacturing curing agents, adhesives, coating materials. It is used in the manufacture of some raw materials for engineering plastics (polyimides) and cross-linking agent for alkyd resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2,4,5-Tetramethylbenzene is the precursor to pyromellitic dianhydride, which is used for manufacturing curing agents, adhesives, coating materials. It is used in the manufacture of some raw materials for engineering plastics (polyimides) and cross-linking agent for alkyd resins.
Solubility
Soluble in alcohol, ether, benzene, and acetone. Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. T. Matsuda.; M. Asanuma.; E. Kikuchi. Effect of high-temperature treatment on the activity of montmorillonite pillared by alumina in the conversion of 1,2,4-trimethylbenzene. Applied Catalysis. 1988, 38 (2), 289-299.
  2. Sadamu Takeda.; Hideaki Chihara. Methyl rotation in 1,2,4,5-tetramethylbenzene and tetramethylpyrazine in the solid state. Tunneling spectra by proton NMR. Journal of Magnetic Resonance (1969). 1983, 54 (2), 285-299.