4-Chloro-2,5-dimethoxyaniline, 98%, Thermo Scientific Chemicals
4-Chloro-2,5-dimethoxyaniline, 98%, Thermo Scientific Chemicals
4-Chloro-2,5-dimethoxyaniline, 98%, Thermo Scientific Chemicals
4-Chloro-2,5-dimethoxyaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloro-2,5-dimethoxyaniline, 98%, Thermo Scientific Chemicals
CAS: 6358-64-1 | C8H10ClNO2 | 187.62 g/mol
Have Questions?
Catalog NumberQuantity
ALFL04202.1425 g
Catalog number ALFL04202.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Chloro-2,5-dimethoxyaniline
CAS6358-64-1
Health Hazard 1H302
Health Hazard 2GHS H Statement
H301-H373
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3P264b-P270-P301+P312-P330-P501c