N-Allylthiourea, 98%, Thermo Scientific Chemicals
N-Allylthiourea, 98%, Thermo Scientific Chemicals
N-Allylthiourea, 98%, Thermo Scientific Chemicals
N-Allylthiourea, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Allylthiourea, 98%, Thermo Scientific Chemicals
CAS: 109-57-9 | C4H8N2S | 116.182 g/mol
Have Questions?
Catalog NumberQuantity
ALFL03377.1425 g
Catalog number ALFL03377.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1768-64-5
IUPAC Name4-chlorooxane
Molecular FormulaC5H9ClO
InChI KeyDHRSKOBIDIDMJZ-UHFFFAOYSA-N
SMILESClC1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Colorless to dark yellow
GC>=95.0 %
Infrared spectrumConforms
N-Allylthiourea is a nitrification inhibitor used in the study on the transformation of diclofenac, naproxen and bisoprolol under aerobic and anaerobic conditions. It is also used in medicine to minimize scar tissue in order to fight against a type of dermatitis. Further, it inhibits the growth of transplanted tumors in mice. It acts as a chelating agent. In addition, it is used in cosmetics, preservative and in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Allylthiourea is a nitrification inhibitor used in the study on the transformation of diclofenac, naproxen and bisoprolol under aerobic and anaerobic conditions. It is also used in medicine to minimize scar tissue in order to fight against a type of dermatitis. Further, it inhibits the growth of transplanted tumors in mice. It acts as a chelating agent. In addition, it is used in cosmetics, preservative and in organic synthesis.
Solubility
Soluble in water and alcohol. Slightly soluble in diethyl ether. Insoluble in benzene.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Lee, W. S.; Chua, A. S. M.; Yeoh, H. K.; Nittami, T.; Ngoh, G. C. Strategy for the biotransformation of fermented palm oil mill effluent into biodegradable polyhydroxyalkanoates by activated sludge. Chem. Eng. J. 2015, 269, 288-297.
  2. Meriac, A.; Tilburg, T. P. A. V.; Eding, E. H.; Kamstra, A.; Schrama, J. W.; Verreth, J. A. J. Effects of diet composition and ultrasound treatment on particle size distribution and carbon bioavailability in feces of rainbow trout. Aquacult. Eng. 2015, 65, 10-16.