8-Methylquinoline, 97+%, Thermo Scientific Chemicals
8-Methylquinoline, 97+%, Thermo Scientific Chemicals
8-Methylquinoline, 97+%, Thermo Scientific Chemicals
8-Methylquinoline, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
8-Methylquinoline, 97+%, Thermo Scientific Chemicals
CAS: 611-32-5 | C10H9N | 143.19 g/mol
Have Questions?
Catalog NumberQuantity
ALFL03288.1425 g
Catalog number ALFL03288.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material8-Methylquinoline
CAS611-32-5
Health Hazard 1H302-H315-H319-H335-H341
Health Hazard 2GHS H Statement
H341-H315-H319
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
View more
8-Methylquinoline has been used in preparation of osmium chloridophosphine complexes, as quinoline carbene tautomers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
8-Methylquinoline has been used in preparation of osmium chloridophosphine complexes, as quinoline carbene tautomers.
Solubility
Soluble in water 0.1-0.5 g/100 mL at 17°C.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Doris Kunz. Synthetic routes to N-heterocyclic carbene complexes: pyridine-carbene tautomerizations. Angewandte Chemie (International Edition). 2007, 46, (19), 3405-3408.
  2. C E Scharping; C C Duke; G M Holder; D Larden. The hepatic metabolism of two methylquinolines. Carcinogenesis. 1993, 14, (5), 1041-1047.