Adonitol, 99%
Adonitol, 99%
Adonitol, 99%
Thermo Scientific Chemicals
Adonitol, 99%
CAS: 488-81-3 | C5H12O5 | 152.146 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFL03253.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAdonitol
CAS488-81-3
Melting Point102°C to 104°C
Recommended StorageAmbient temperatures; Store under Nitrogen
RTECS NumberVJ0800000
View more
Adonitol is metabolized to teicholic acids used in the cell walls of gram positive bacteria. Adonitol is often compared to other cell permeating molecules such as formamide, propanediol, and DMSO as a cryopreservation agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Adonitol is metabolized to teicholic acids used in the cell walls of gram positive bacteria. Adonitol is often compared to other cell permeating molecules such as formamide, propanediol, and DMSO as a cryopreservation agent.
Solubility
Soluble in water (100 mg/ml).
Notes
Hygroscopic. Store under dry inert gas. Incompatible with water and oxidizing agents.
RUO – Research Use Only
General References:
  1. Graciela F. DE Valdéz; Graciela S. DE Giori; Aída A. P. DE Ruiz Holgado and Guillermo Oliver. Protective Effect of Adonitol on Lactic Acid Bacteria Subjected to Freeze-Drying.Appl. Environ. Microbiol.1983, 45 302-304.
  2. A. G. Fadnis; S. K. Kulshrestha. Kinetics of oxidation of adonitol by manganese(III) pyrophosphate.Reaction Kinetics and Catalysis Letters.1982, 19 267-269.