2-Ethyltoluene, 98+%, Thermo Scientific Chemicals
2-Ethyltoluene, 98+%, Thermo Scientific Chemicals
2-Ethyltoluene, 98+%, Thermo Scientific Chemicals
2-Ethyltoluene, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Ethyltoluene, 98+%, Thermo Scientific Chemicals
CAS: 611-14-3 | C9H12 | 120.195 g/mol
Have Questions?
Catalog NumberQuantity
ALFL02028.065 g
Catalog number ALFL02028.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS937-14-4
IUPAC Name3-chlorobenzene-1-carboperoxoic acid
Molecular FormulaC7H5ClO3
InChI KeyNHQDETIJWKXCTC-UHFFFAOYSA-N
SMILESOOC(=O)C1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Moist powder
3-Chlorobenzoic acid=<12 %
HPLCdi-m-Chlorobenzoyl peroxide: < 0.1% Typical
HPLC=<0.2 % di-m-Chlorobenzoyl peroxide
Titration Iodometric70 to 75 %
View more
2-Ethyltoluene, is used to predict novel lead compounds for drug discovery that depends on the quality of the docking scoring function.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Ethyltoluene, is used to predict novel lead compounds for drug discovery that depends on the quality of the docking scoring function.
Solubility
Soluble in water (74.6 mg/L (25°C)).
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Monica Alonso.; Laura Cerdan.; Anna Godayol.; Enriqueta Antico.; Juan M. Sanchez. Headspace needle-trap analysis of priority volatile organic compounds from aqueous samples: Application to the analysis of natural and waste waters. Journal of Chromatography A. 2011, 1218, (45), 8131-8139.
  2. A. Roubaud.; R. Minetti.; L.R. Sochet. Oxidation and combustion of low alkylbenzenes at high pressure: comparative reactivity and auto-ignition. Combustion and Flame. 2000, 121, (3), 535-541.