3-Thiophenemethanol, 97%, Thermo Scientific Chemicals
3-Thiophenemethanol, 97%, Thermo Scientific Chemicals
3-Thiophenemethanol, 97%, Thermo Scientific Chemicals
3-Thiophenemethanol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Thiophenemethanol, 97%, Thermo Scientific Chemicals
CAS: 71637-34-8 | C5H6OS | 114.16 g/mol
Have Questions?
Catalog NumberQuantity
ALFL01935.065 g
Catalog number ALFL01935.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7718-98-1
IUPAC Namevanadium(3+) trichloride
Molecular FormulaCl3V
InChI KeyHQYCOEXWFMFWLR-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[V+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=96.0 %
Appearance (Form)Powder
Insoluble matter=<0.5 % (in water)
Appearance (Color)Dark blue to violet to black
3-Thiophenemethanol was used in the preparation of 3-substituted thiophene conducting copolymers which has potential applications in electrochromic displays.1 It was also used in the synthesis of 4-(thiophene-3-ylmethoxy)phthalonitrile
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Thiophenemethanol was used in the preparation of 3-substituted thiophene conducting copolymers which has potential applications in electrochromic displays.1 It was also used in the synthesis of 4-(thiophene-3-ylmethoxy)phthalonitrile
Solubility
Slightly soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only