2-Ethylphenol, 98+%, Thermo Scientific Chemicals
2-Ethylphenol, 98+%, Thermo Scientific Chemicals
2-Ethylphenol, 98+%, Thermo Scientific Chemicals
2-Ethylphenol, 98+%, Thermo Scientific Chemicals
2-Ethylphenol, 98+%, Thermo Scientific Chemicals
CAS: 90-00-6 | C8H10O | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number ALFL01637.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Ethylphenol
CAS90-00-6
Health Hazard 1H227-H302+H312+H332-H314
Health Hazard 2GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
Health Hazard 3P210-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c