3-Iodoaniline, 98%, Thermo Scientific Chemicals
3-Iodoaniline, 98%, Thermo Scientific Chemicals
3-Iodoaniline, 98%, Thermo Scientific Chemicals
3-Iodoaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Iodoaniline, 98%, Thermo Scientific Chemicals
CAS: 626-01-7 | C6H6IN | 219.025 g/mol
Have Questions?
Catalog NumberQuantity
ALFL01159.0910 g
Catalog number ALFL01159.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS329-15-7
IUPAC Name4-(trifluoromethyl)benzoyl chloride
Molecular FormulaC8H4ClF3O
InChI KeyOXZYBOLWRXENKT-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(C=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless to yellow
Refractive index1.4755 to 1.4775 (20°C, 589 nm)
Infrared spectrumConforms
3-Iodoanilineis used as Anilines, Aromatic Amines and Nitro Compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Iodoanilineis used as Anilines, Aromatic Amines and Nitro Compounds.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store at 4°C. Store protected from light. Incompatible with Acid chlorides, acid anhydrides, acids, chloroformates, strong oxidizing agents.
RUO – Research Use Only