p-Toluenesulfonic anhydride, 97%, Thermo Scientific Chemicals
p-Toluenesulfonic anhydride, 97%, Thermo Scientific Chemicals
p-Toluenesulfonic anhydride, 97%, Thermo Scientific Chemicals
p-Toluenesulfonic anhydride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenesulfonic anhydride, 97%, Thermo Scientific Chemicals
CAS: 4124-41-8 | C14H14O5S2 | 326.381 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFL00160.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS533-31-3
IUPAC Name2H-1,3-benzodioxol-5-ol
Molecular FormulaC7H6O3
InChI KeyLUSZGTFNYDARNI-UHFFFAOYSA-N
SMILESOC1=CC=C2OCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Crystalline powder
Water=<0.2 % (K.F.)
Appearance (Color)Off-white to beige