Nystatin, 10 mg/ml in methanol, sterile-filtered, Thermo Scientific Chemicals
Nystatin, 10 mg/ml in methanol, sterile-filtered, Thermo Scientific Chemicals
Nystatin, 10 mg/ml in methanol, sterile-filtered, Thermo Scientific Chemicals
Nystatin, 10 mg/ml in methanol, sterile-filtered, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nystatin, 10 mg/ml in methanol, sterile-filtered, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFJ67369.XF1 mL
Catalog number ALFJ67369.XF
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialNystatin
TypeNystatin
Physical FormLiquid
CAS1400-61-9
Health Hazard 1H225-H301+H311+H331-H370